Tisk…

Methyl (2E)-3-(2-fluorophenyl)prop-2-enoate 95%

Dodavatel: Apollo Scientific

Synonyms: (E)-methyl 3-(2-fluorophenyl)acrylate

PC28126-1G PC28126-5G
APOSPC28126-1GEA 5360 CZK
APOSPC28126-1G APOSPC28126-5G
Methyl (2E)-3-(2-fluorophenyl)prop-2-enoate 95%
Methyl (2E)-3-(2-fluorophenyl)prop-2-enoate
Vzorec: C₁₀H₉FO₂
MW: 180,176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03102529
CAS číslo: 104201-65-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR