Tisk…

6-Chloro-3-(trifluoromethyl)-[1,2,4]triazolo[4,3-b]pyridazine 95%

Dodavatel: Apollo Scientific
Varování

PC28069-1G PC28069-250MG PC28069-5G
APOSPC28069-1GEA 9100 CZK
APOSPC28069-1G APOSPC28069-250MG APOSPC28069-5G
6-Chloro-3-(trifluoromethyl)-[1,2,4]triazolo[4,3-b]pyridazine 95%
6-Chloro-3-(trifluoromethyl)-[1,2,4]triazolo[4,3-b]pyridazine
Vzorec: C₆H₂ClF₃N₄
MW: 222,555 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00505462
CAS číslo: 40971-95-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR