Tisk…

4-Bromo-2,6-difluorobenzyl fluoride 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC27936-250MG
APOSPC27936-250MGEA 2280 CZK
APOSPC27936-250MG
4-Bromo-2,6-difluorobenzyl fluoride 95%
4-Bromo-2,6-difluorobenzyl fluoride
Vzorec: C₇H₄BrF₃
MW: 225,006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31664910
CAS číslo: 2244087-29-2
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR