Tisk…

2-Bromo-5-fluoro-3-nitrobenzyl fluoride 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC27920-1G PC27920-250MG
APOSPC27920-1GEA 17360 CZK
APOSPC27920-1G APOSPC27920-250MG
2-Bromo-5-fluoro-3-nitrobenzyl fluoride 95%
2-Bromo-5-fluoro-3-nitrobenzyl fluoride
Vzorec: C₇H₄BrF₂NO₂
MW: 252,013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28738751
CAS číslo: 1807004-68-7
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR