5-Bromo-2-iodobenzyl fluoride 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC27894-250MG
APOSPC27894-250MGEA
2280
CZK
APOSPC27894-250MG
5-Bromo-2-iodobenzyl fluoride 95%
5-Bromo-2-iodobenzyl fluoride
Vzorec:
C₇H₅BrFI MW: 314,921 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31664892 CAS číslo: 2244088-43-3 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...