Tisk…

Fmoc-D-2,4,5-Trifluorophenylalanine

Dodavatel: Apollo Scientific

PC27884-1G PC27884-250MG
APOSPC27884-1GEA 11610 CZK
APOSPC27884-1G APOSPC27884-250MG
Fmoc-D-2,4,5-Trifluorophenylalanine
Fmoc-D-2,4,5-Trifluorophenylalanine
Vzorec: C₂₄H₁₈F₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD06659134
CAS číslo: 1217837-13-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR