Tisk…

1-([(9H-Fluoren-9-ylmethoxy)carbonyl]amino)-4,4-difluorocyclohexane-1-carboxylic acid

Dodavatel: Apollo Scientific

PC27868-100MG PC27868-250MG
APOSPC27868-100MGEA 22780 CZK
APOSPC27868-100MG APOSPC27868-250MG
1-([(9H-Fluoren-9-ylmethoxy)carbonyl]amino)-4,4-difluorocyclohexane-1-carboxylic acid
1-([(9H-Fluoren-9-ylmethoxy)carbonyl]amino)-4,4-difluorocyclohexane-1-carboxylic acid
Vzorec: C₂₂H₂₁F₂NO₄
MW: 401,403 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28389033
CAS číslo: 1986905-26-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR