Tisk…

(2,4-Difluoro-phenyl)-[(9H-fluoren-9-ylmethoxycarbonylamino)]-acetic acid

Dodavatel: Apollo Scientific

PC27855-100MG PC27855-1G PC27855-250MG
APOSPC27855-100MGEA 14520 CZK
APOSPC27855-100MG APOSPC27855-1G APOSPC27855-250MG
(2,4-Difluoro-phenyl)-[(9H-fluoren-9-ylmethoxycarbonylamino)]-acetic acid
(2,4-Difluoro-phenyl)-[(9H-fluoren-9-ylmethoxycarbonylamino)]-acetic acid
Vzorec: C₂₃H₁₇F₂NO₄
MW: 409,382 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09026220
CAS číslo: 678991-01-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR