Tisk…

4-Bromo-2-fluorophenylhydrazine hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromo-2-fluoro-1-hydrazinobenzene hydrochloride, (4-Bromo-2-fluorophenyl)hydrazine hydrochloride

PC2715-25G PC2715-5G PC2715-100G
APOSPC2715-25GEA 1240 CZK
APOSPC2715-25G APOSPC2715-5G APOSPC2715-100G
4-Bromo-2-fluorophenylhydrazine hydrochloride
4-Bromo-2-fluorophenylhydrazine hydrochloride
Vzorec: C₆H₆BrFN₂·ClH
MW: 241,489 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03094166
CAS číslo: 502496-24-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR