3-[2-(Trifluoromethoxy)phenyl]propanoic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(Trifluoromethoxy)hydrocinnamic acid, 2-(2-Carboxyethyl)-α,α,α-trifluoroanisole, 3-(2-(Trifluoromethoxy)phenyl)propanoic acid
PC2638-1G
PC2638-250MG
APOSPC2638-1GEA
7050
CZK
APOSPC2638-1G
APOSPC2638-250MG
3-[2-(Trifluoromethoxy)phenyl]propanoic acid
3-[2-(Trifluoromethoxy)phenyl]propanoic acid
Vzorec:
C₁₀H₉F₃O₃ MW: 234,172 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08458088 CAS číslo: 914636-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...