4-Iodo-β,β,β-trifluorophenetole
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Iodo-4-(2,2,2-trifluoroethoxy)benzene, 4-Iodophenyl 2,2,2-trifluoroethyl ether
PC2634-1G
PC2634-5G
APOSPC2634-1GEA
3610
CZK
APOSPC2634-1G
APOSPC2634-5G
4-Iodo-β,β,β-trifluorophenetole
4-Iodo-β,β,β-trifluorophenetole
Vzorec:
C₈H₆F₃IO Storage Temperature: Ambient |
MDL Number:
MFCD08458086 CAS číslo: 530080-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...