4-Fluoro-3-(methylsulphonyl)aniline 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Amino-2-fluorophenyl methyl sulphone
PC2626-1G
PC2626-5G
APOSPC2626-1GEA
3540
CZK
APOSPC2626-1G
APOSPC2626-5G
4-Fluoro-3-(methylsulphonyl)aniline 98%
4-Fluoro-3-(methylsulphonyl)aniline
Vzorec:
C₇H₈FNO₂S MW: 189.207 g/mol Bod tání: 99…101 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08458080 CAS číslo: 914636-44-5 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...