2-Fluoro-5-nitrophenyl methyl sulphone
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Fluoro-2-(methylsulphonyl)-4-nitrobenzene, 4-Fluoro-3-(methylsulphonyl)nitrobenzene
PC2625-1G
PC2625-5G
APOSPC2625-1GEA
1120
CZK
APOSPC2625-1G
APOSPC2625-5G
2-Fluoro-5-nitrophenyl methyl sulphone
2-Fluoro-5-nitrophenyl methyl sulphone
Vzorec:
C₇H₆FNO₄S MW: 219.19 g/mol Bod tání: 128…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD08458079 CAS číslo: 914636-41-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...