Tisk…

2-(Hydroxymethyl)-5-(trifluoromethyl)furan 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: [5-(Trifluoromethyl)fur-2-yl]methanol, 5-(Trifluoromethyl)furfuryl alcohol

PC2621-1G PC2621-250MG
APOSPC2621-1GEA 13460 CZK
APOSPC2621-1G APOSPC2621-250MG
2-(Hydroxymethyl)-5-(trifluoromethyl)furan 95%
2-(Hydroxymethyl)-5-(trifluoromethyl)furan
Vzorec: C₆H₅F₃O₂
MW: 166,098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08060542
CAS číslo: 65865-28-3
UN: 1986
ADR: 3+6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR