Tisk…

3,4-Difluoropyridine

Dodavatel: Apollo Scientific
Nebezpečí

PC2502-1G PC2502-250MG
APOSPC2502-1GEA 12070 CZK
APOSPC2502-1G APOSPC2502-250MG
3,4-Difluoropyridine
3,4-Difluoropyridine
Vzorec: C₅H₃F₂N
MW: 115,081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114246
CAS číslo: 82878-63-5
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR