Tisk…

Fluoromethyl 4-methylbenzenesulphonate 97+%

Dodavatel: Apollo Scientific
Nebezpečí

PC250002-1G
APOSPC250002-1GEA 2110 CZK
APOSPC250002-1G
Fluoromethyl 4-methylbenzenesulphonate 97+%
Fluoromethyl 4-methylbenzenesulphonate
Vzorec: C₈H₉FO₃S
MW: 204,219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20482732
CAS číslo: 114435-86-8
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR