Tisk…

1H,1H-Nonacosafluoro(2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecan-1-ol) 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-Heptadecafluoro-2,5,8,11-tetrakis(trifluoromethyl)-3,6,9,12-tetraoxapentadecan-1-ol

PC2481-25G PC2481-5G
APOSPC2481-25GEA 0 CZK
APOSPC2481-25G APOSPC2481-5G
1H,1H-Nonacosafluoro(2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecan-1-ol) 97%
1H,1H-Nonacosafluoro(2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecan-1-ol)
Vzorec: C₁₅H₃F₂₉O₅
MW: 814,135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460480
CAS číslo: 141977-66-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR