Tisk…

4'-Bromo-4-(trifluoromethoxy)biphenyl 98%

Dodavatel: Apollo Scientific
Varování

PC2475-1G PC2475-250MG
APOSPC2475-1GEA 11410 CZK
APOSPC2475-1G APOSPC2475-250MG
4'-Bromo-4-(trifluoromethoxy)biphenyl 98%
4'-Bromo-4-(trifluoromethoxy)biphenyl
Vzorec: C₁₃H₈BrF₃O
MW: 317.101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972698
CAS číslo: 134150-03-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR