Perfluoro-1,9-dibromo-2,5,8-trioxanonane
Dodavatel: Apollo Scientific
|
Varování
|
PC2472-25G
PC2472-5G
PC2472-1G
APOSPC2472-25GEA
11180
CZK
APOSPC2472-25G
APOSPC2472-5G
APOSPC2472-1G
Perfluoro-1,9-dibromo-2,5,8-trioxanonane
Perfluoro-1,9-dibromo-2,5,8-trioxanonane
Vzorec:
C₆Br₂F₁₂O₃ Bod varu: 52 °C (25 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD03094159 CAS číslo: 330562-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...