Tisk…

2,4-Dimethyl-3-fluorobromobenzene

Dodavatel: Apollo Scientific

Synonyms: 2,4-Dimethyl-3-fluoro-bromobenzene

PC2447-1G PC2447-500MG PC2447-5G
APOSPC2447-1GEA 8870 CZK
APOSPC2447-1G APOSPC2447-500MG APOSPC2447-5G
2,4-Dimethyl-3-fluorobromobenzene
2,4-Dimethyl-3-fluorobromobenzene
Vzorec: C₈H₈BrF
MW: 203,052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079714
CAS číslo: 26584-26-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR