Tisk…

2-Bromo-N,N-dimethyl-4-fluoroaniline 98%

Dodavatel: Apollo Scientific
Varování

PC2386-1G PC2386-5G
APOSPC2386-1GEA 4960 CZK
APOSPC2386-1G APOSPC2386-5G
2-Bromo-N,N-dimethyl-4-fluoroaniline 98%
2-Bromo-N,N-dimethyl-4-fluoroaniline
Vzorec: C₈H₉BrFN
Storage Temperature: Refrigerator
MDL Number: MFCD04116317
CAS číslo: 887268-19-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR