3,4-Bis(trifluoromethyl)benzoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC2239-1G
PC2239-250MG
APOSPC2239-1GEA
5960
CZK
APOSPC2239-1G
APOSPC2239-250MG
3,4-Bis(trifluoromethyl)benzoic acid 98%
3,4-Bis(trifluoromethyl)benzoic acid
Vzorec:
C₉H₄F₆O₂ MW: 258,117 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07779330 CAS číslo: 133804-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...