3-(4-Fluorophenylthio)butyric acid
Dodavatel: Apollo Scientific
PC2189-1G
PC2189-500MG
APOSPC2189-1GEA
13260
CZK
APOSPC2189-1G
APOSPC2189-500MG
3-(4-Fluorophenylthio)butyric acid
3-(4-Fluorophenylthio)butyric acid
Vzorec:
C₁₀H₁₁FO₂S Storage Temperature: Ambient |
MDL Number:
MFCD00816765 CAS číslo: 259544-93-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...