N-(2-Fluoroethyl)aniline ≥98%
Dodavatel: Apollo Scientific
PC210144-100MG
PC210144-1G
PC210144-250MG
APOSPC210144-100MGEA
4770
CZK
APOSPC210144-100MG
APOSPC210144-1G
APOSPC210144-250MG
N-(2-Fluoroethyl)aniline ≥98%
N-(2-Fluoroethyl)aniline
Vzorec:
C₈H₁₀FN Storage Temperature: Ambient |
MDL Number:
MFCD19217349 CAS číslo: 459-40-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...