Tisk…

Ethyl 6-fluoro-4-hydroxy-2-naphthoate ≥98%

Dodavatel: Apollo Scientific

PC210131-1G PC210131-250MG
APOSPC210131-1GEA 30060 CZK
APOSPC210131-1G APOSPC210131-250MG
Ethyl 6-fluoro-4-hydroxy-2-naphthoate ≥98%
Ethyl 6-fluoro-4-hydroxy-2-naphthoate
Vzorec: C₁₃H₁₁FO₃
MW: 234,223 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181239
CAS číslo: 1093073-40-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR