3-(Difluoromethoxy)cyclobutane-1-carboxylic acid ≥98%
Dodavatel: Apollo Scientific
PC210130-100MG
PC210130-250MG
APOSPC210130-100MGEA
13560
CZK
APOSPC210130-100MG
APOSPC210130-250MG
3-(Difluoromethoxy)cyclobutane-1-carboxylic acid ≥98%
3-(Difluoromethoxy)cyclobutane-1-carboxylic acid
Vzorec:
C₆H₈F₂O₃ MW: 166,123 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28383706 CAS číslo: 1803603-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...