Tisk…

1-(Cyclopropyloxy)-3,5-difluorobenzene ≥98%

Dodavatel: Apollo Scientific

Synonyms: 1-Cyclopropoxy-3,5-difluorobenzene

PC210129-100MG PC210129-1G PC210129-250MG
APOSPC210129-100MGEA 4800 CZK
APOSPC210129-100MG APOSPC210129-1G APOSPC210129-250MG
1-(Cyclopropyloxy)-3,5-difluorobenzene ≥98%
1-(Cyclopropyloxy)-3,5-difluorobenzene
Vzorec: C₉H₈F₂O
MW: 170,156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16996191
CAS číslo: 1138331-79-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR