Tisk…

5-Fluoro-6-methoxy-2,3-dihydro-1H-inden-1-one

Dodavatel: Apollo Scientific

PC210040-1G PC210040-500MG
APOSPC210040-1GEA 15310 CZK
APOSPC210040-1G APOSPC210040-500MG
5-Fluoro-6-methoxy-2,3-dihydro-1H-inden-1-one
5-Fluoro-6-methoxy-2,3-dihydro-1H-inden-1-one
Vzorec: C₁₀H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD09261202
CAS číslo: 83802-71-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR