Tetrabutylazanium trifluoro(4-fluorophenyl)boranuide
Dodavatel: Apollo Scientific
PC210021-1G
PC210021-5G
APOSPC210021-1GEA
2900
CZK
APOSPC210021-1G
APOSPC210021-5G
Tetrabutylazanium trifluoro(4-fluorophenyl)boranuide
Tetrabutylazanium trifluoro(4-fluorophenyl)boranuide
Vzorec:
C₆H₄BF₄·N(C₄H₉)₄ MW: 405,364 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28101518 CAS číslo: 1291068-40-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...