Chromium(III) trifluoroacetate 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC2078E-5G
PC2078E-1G
APOSPC2078E-5GEA
4100
CZK
APOSPC2078E-5G
APOSPC2078E-1G
Chromium(III) trifluoroacetate 97%
Chromium(III) trifluoroacetate
Vzorec:
Cr(CF₃CO₂)₃ Storage Temperature: Ambient |
MDL Number:
MFCD00069078 CAS číslo: 16712-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...