Tisk…

(Z)-1,1,1,3-Tetrafluorobut-2-ene

Dodavatel: Apollo Scientific

PC2071-25G PC2071-5G
APOSPC2071-25GEA 25430 CZK
APOSPC2071-25G APOSPC2071-5G
(Z)-1,1,1,3-Tetrafluorobut-2-ene
(Z)-1,1,1,3-Tetrafluorobut-2-ene
Vzorec: C₄H₄F₄
MW: 128,068 g/mol
Bod varu: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD07784203
CAS číslo: 791616-88-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR