2-Chloro-4-fluoronicotinic acid 97%
Dodavatel: Apollo Scientific
PC201376-1G
PC201376-5G
APOSPC201376-1GEA
2440
CZK
APOSPC201376-1G
APOSPC201376-5G
2-Chloro-4-fluoronicotinic acid 97%
2-Chloro-4-fluoronicotinic acid
Vzorec:
C₆H₃ClFNO₂ |
MDL Number:
MFCD11040264 CAS číslo: 929022-76-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...