4-Fluoro-6-iodo-1H-indazole 95+%
Dodavatel: Apollo Scientific
PC201360-100MG
PC201360-250MG
PC201360-1G
APOSPC201360-100MGEA
6050
CZK
APOSPC201360-100MG
APOSPC201360-250MG
APOSPC201360-1G
4-Fluoro-6-iodo-1H-indazole 95+%
4-Fluoro-6-iodo-1H-indazole
Vzorec:
C₇H₄FIN₂ MW: 262,023 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07781847 CAS číslo: 887568-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...