Tisk…

4-(4-(Trifluoromethyl)phenyl)oxazol-2-amine 96%

Dodavatel: Apollo Scientific

PC201327-100MG PC201327-250MG
APOSPC201327-100MGEA 2780 CZK
APOSPC201327-100MG APOSPC201327-250MG
4-(4-(Trifluoromethyl)phenyl)oxazol-2-amine 96%
4-(4-(Trifluoromethyl)phenyl)oxazol-2-amine
Vzorec: C₁₀H₇F₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD14706776
CAS číslo: 859721-53-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR