4-Bromo-2-(difluoromethoxy)pyridine 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC201316-100MG
PC201316-250MG
APOSPC201316-100MGEA
2330
CZK
APOSPC201316-100MG
APOSPC201316-250MG
4-Bromo-2-(difluoromethoxy)pyridine 97%
4-Bromo-2-(difluoromethoxy)pyridine
Vzorec:
C₆H₄BrF₂NO MW: 224,003 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14584703 CAS číslo: 832735-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...