Tisk…

3,5-Difluoro-4-formylbenzoic acid 97%

Dodavatel: Apollo Scientific

Synonyms: 4-Carboxy-2,6-difluorobenzaldehyde

PC201295-250MG PC201295-1G PC201295-5G
APOSPC201295-250MGEA 2370 CZK
APOSPC201295-250MG APOSPC201295-1G APOSPC201295-5G
3,5-Difluoro-4-formylbenzoic acid 97%
3,5-Difluoro-4-formylbenzoic acid
Vzorec: C₈H₄F₂O₃
MW: 186.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835188
CAS číslo: 736990-88-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR