Tisk…

5-Bromo-8-fluoroisoquinoline 98%

Dodavatel: Apollo Scientific

PC201279-1G PC201279-5G
APOSPC201279-1GEA 9030 CZK
APOSPC201279-1G APOSPC201279-5G
5-Bromo-8-fluoroisoquinoline 98%
5-Bromo-8-fluoroisoquinoline
Vzorec: C₉H₅BrFN
MW: 226,045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18447885
CAS číslo: 679433-94-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR