Ethyl 5-bromo-2-fluorobenzoate 95+%
Dodavatel: Apollo Scientific
PC201270-10G
PC201270-1G
PC201270-5G
APOSPC201270-10GEA
9790
CZK
APOSPC201270-10G
APOSPC201270-1G
APOSPC201270-5G
Ethyl 5-bromo-2-fluorobenzoate 95+%
Ethyl 5-bromo-2-fluorobenzoate
Vzorec:
C₉H₈BrFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD06204416 CAS číslo: 612835-53-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...