Tisk…

(S)-4,4-Difluoropyrrolidine-2-carboxylic acid 95+%

Dodavatel: Apollo Scientific

PC201261-250MG PC201261-1G
APOSPC201261-250MGEA 2120 CZK
APOSPC201261-250MG APOSPC201261-1G
(S)-4,4-Difluoropyrrolidine-2-carboxylic acid 95+%
(S)-4,4-Difluoropyrrolidine-2-carboxylic acid
Vzorec: C₅H₇F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD10697866
CAS číslo: 52683-81-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR