6-Chloro-3-(trifluoromethyl)picolinonitrile 95+%
Dodavatel: Apollo Scientific
PC201249-250MG
PC201249-1G
PC201249-5G
APOSPC201249-250MGEA
2320
CZK
APOSPC201249-250MG
APOSPC201249-1G
APOSPC201249-5G
6-Chloro-3-(trifluoromethyl)picolinonitrile 95+%
6-Chloro-3-(trifluoromethyl)picolinonitrile
Vzorec:
C₇H₂ClF₃N₂ MW: 206,552 g/mol |
MDL Number:
MFCD18261725 CAS číslo: 401590-41-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...