5-(Benzyloxy)-2-fluoroaniline 95+%
Dodavatel: Apollo Scientific
PC201234-1G
PC201234-250MG
PC201234-5G
APOSPC201234-1GEA
4730
CZK
APOSPC201234-1G
APOSPC201234-250MG
APOSPC201234-5G
5-(Benzyloxy)-2-fluoroaniline 95+%
5-(Benzyloxy)-2-fluoroaniline
Vzorec:
C₁₃H₁₂FNO MW: 217,239 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00835609 CAS číslo: 342042-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...