Tisk…

(S)-2-(4-Fluorophenyl)pyrrolidine 98%

Dodavatel: Apollo Scientific

PC201224-100MG PC201224-1G PC201224-250MG
APOSPC201224-100MGEA 4200 CZK
APOSPC201224-100MG APOSPC201224-1G APOSPC201224-250MG
(S)-2-(4-Fluorophenyl)pyrrolidine 98%
(S)-2-(4-Fluorophenyl)pyrrolidine
Vzorec: C₁₀H₁₂FN
MW: 165,207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06761856
CAS číslo: 298690-90-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR