Tisk…

5-Fluoro-3,4-dihydroisoquinolin-1(2H)-one 98%

Dodavatel: Apollo Scientific

PC201205-1G PC201205-250MG
APOSPC201205-1GEA 5720 CZK
APOSPC201205-1G APOSPC201205-250MG
5-Fluoro-3,4-dihydroisoquinolin-1(2H)-one 98%
5-Fluoro-3,4-dihydroisoquinolin-1(2H)-one
Vzorec: C₉H₈FNO
MW: 165,164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114877
CAS číslo: 230301-83-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR