1-Chloro-6-fluoroisoquinoline 95+%
Dodavatel: Apollo Scientific
PC201195-100MG
PC201195-1G
PC201195-250MG
APOSPC201195-100MGEA
2860
CZK
APOSPC201195-100MG
APOSPC201195-1G
APOSPC201195-250MG
1-Chloro-6-fluoroisoquinoline 95+%
1-Chloro-6-fluoroisoquinoline
Vzorec:
C₉H₅ClFN MW: 181,594 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09959717 CAS číslo: 214045-86-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...