Tisk…

2-(Trifluoromethyl)benzo[d]oxazole 95+%

Dodavatel: Apollo Scientific

PC201191-1G PC201191-250MG PC201191-5G
APOSPC201191-1GEA 8630 CZK
APOSPC201191-1G APOSPC201191-250MG APOSPC201191-5G
2-(Trifluoromethyl)benzo[d]oxazole 95+%
2-(Trifluoromethyl)benzo[d]oxazole
Vzorec: C₈H₄F₃NO
MW: 187,119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00480774
CAS číslo: 2008-04-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR