Tisk…

3-Fluoroquinoline-6-carboxylic acid 95+%

Dodavatel: Apollo Scientific

PC201183-100MG PC201183-250MG PC201183-25MG
APOSPC201183-100MGEA 10190 CZK
APOSPC201183-100MG APOSPC201183-250MG APOSPC201183-25MG
3-Fluoroquinoline-6-carboxylic acid 95+%
3-Fluoroquinoline-6-carboxylic acid
Vzorec: C₁₀H₆FNO₂
MW: 191.159 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28133464
CAS číslo: 1824275-93-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR