4-Chloro-5-fluoro-2-methoxypyrimidine 97%
Dodavatel: Apollo Scientific
PC201178-1G
PC201178-250MG
PC201178-5G
APOSPC201178-1GEA
3380
CZK
APOSPC201178-1G
APOSPC201178-250MG
APOSPC201178-5G
4-Chloro-5-fluoro-2-methoxypyrimidine 97%
4-Chloro-5-fluoro-2-methoxypyrimidine
Vzorec:
C₅H₄ClFN₂O Storage Temperature: Ambient |
MDL Number:
MFCD09834963 CAS číslo: 1801-06-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...