Tisk…

4-Fluorobenzo[d]isoxazol-3(2H)-one 98%

Dodavatel: Apollo Scientific

PC201177-1G
APOSPC201177-1GEA 3380 CZK
APOSPC201177-1G
4-Fluorobenzo[d]isoxazol-3(2H)-one 98%
4-Fluorobenzo[d]isoxazol-3(2H)-one
Vzorec: C₇H₄FNO₂
MW: 153,111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18804910
CAS číslo: 178747-83-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR