Tisk…

4-Ethoxy-2-fluoropyridine 97%

Dodavatel: Apollo Scientific

PC201174-100MG PC201174-1G PC201174-250MG
APOSPC201174-100MGEA 787 CZK
APOSPC201174-100MG APOSPC201174-1G APOSPC201174-250MG
4-Ethoxy-2-fluoropyridine 97%
4-Ethoxy-2-fluoropyridine
Vzorec: C₇H₈FNO
MW: 141,143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835274
CAS číslo: 175965-82-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR