4-Chloro-5-fluoro-1H-indole 97%
Dodavatel: Apollo Scientific
PC201168-100MG
PC201168-250MG
APOSPC201168-100MGEA
2480
CZK
APOSPC201168-100MG
APOSPC201168-250MG
4-Chloro-5-fluoro-1H-indole 97%
4-Chloro-5-fluoro-1H-indole
Vzorec:
C₈H₅ClFN MW: 169,583 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12924664 CAS číslo: 169674-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...